missing translation for 'onlineSavingsMsg'
Learn More

Florasulam, TRC

Product Code. 30138496
Change view
Click to view available options
Quantity:
50 mg
500 mg
5 g
Unit Size:
500mg
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138496 500 mg 500mg
17972615 5 g 5g
17951395 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138496 Supplier Toronto Research Chemicals Supplier No. TRCF405730500MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145701-23-1
Molecular Formula C12 H8 F3 N5 O3 S
Molecular Weight (g/mol) 359.28
Synonym N-(2,6-Difluorophenyl)-8-fluoro-5-methoxy[1,2,4]triazolo[1,5-c]pyrimidine-2-sulfonamide, Florasulam, Kantor, PrePass XC, Primus, Primus SC
IUPAC Name N-(2,6-difluorophenyl)-8-fluoro-5-methoxy-[1,2,4]triazolo[1,5-c]pyrimidine-2-sulfonamide
SMILES COc1ncc(F)c2nc(nn12)S(=O)(=O)Nc3c(F)cccc3F

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C12H8F3N5O3S/c1-23-12-16-5-8(15)10-17-11(18-20(10)12)24(21,22)19-9-6(13)3-2-4-7(9)14/h2-5,19H,1H3
Formula Weight 359.03
Chemical Name or Material Florasulam
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.