missing translation for 'onlineSavingsMsg'
Learn More

Florasulam, TRC

Product Code. 17972615
Change view
Click to view available options
Quantity:
50 mg
500 mg
5 g
Unit Size:
500mg
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17972615 5 g 5g
30138496 500 mg 500mg
17951395 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17972615 Supplier Toronto Research Chemicals Supplier No. TRCF4057305G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145701-23-1
Formula Weight 359.03
InChI Formula InChI=1S/C12H8F3N5O3S/c1-23-12-16-5-8(15)10-17-11(18-20(10)12)24(21,22)19-9-6(13)3-2-4-7(9)14/h2-5,19H,1H3
IUPAC Name N-(2,6-difluorophenyl)-8-fluoro-5-methoxy-[1,2,4]triazolo[1,5-c]pyrimidine-2-sulfonamide
Molecular Formula C12 H8 F3 N5 O3 S
Molecular Weight (g/mol) 359.28
SMILES COc1ncc(F)c2nc(nn12)S(=O)(=O)Nc3c(F)cccc3F
Synonym N-(2,6-Difluorophenyl)-8-fluoro-5-methoxy[1,2,4]triazolo[1,5-c]pyrimidine-2-sulfonamide, Florasulam, Kantor, PrePass XC, Primus, Primus SC
Chemical Name or Material Florasulam

Specifications

Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.