missing translation for 'onlineSavingsMsg'
Learn More

Flubendazole, 98%

Product Code. 15522283 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15522283 1 g 1g
15532283 5 g 5g
15542283 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15522283 Supplier Thermo Scientific Acros Supplier No. 101910010
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 31430-15-6 | C16H12FN3O3 | 313.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31430-15-6
Molecular Formula C16H12FN3O3
Molecular Weight (g/mol) 313.28
InChI Key CPEUVMUXAHMANV-UHFFFAOYSA-N
Synonym flubendazole, flumoxane, flubendazol, fluvermal, flubenol, flubendazolum, flumoxal, flutelmium, methyl 5-4-fluorobenzoyl-1h-benzo d imidazol-2-yl carbamate, flubendazol inn-spanish
PubChem CID 35802
ChEBI CHEBI:77095
IUPAC Name methyl N-[6-(4-fluorobenzoyl)-1H-benzimidazol-2-yl]carbamate
SMILES COC(=O)NC1=NC2=C(N1)C=C(C=C2)C(=O)C3=CC=C(C=C3)F

Specifications

Melting Point 260.0°C
Loss on Drying 0.5% max. (1 g, 105°C, 4 hr)
Infrared Spectrum Authentic
Quantity 1 g
Formula Weight 313.28
Chemical Name or Material Flubendazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.