missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flubendiamide 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Flubendiamide |
| CAS | 272451-65-7 |
| Analyte or Component Names | Flubendiamide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H22 F7 I N2 O4 S |
| Concentration | 100 μg/mL |
| Certifications/Compliance | ISO 17034 |
| InChI Formula | InChI=1S/C23H22F7IN2O4S/c1-12-10-13(21(24,22(25,26)27)23(28,29)30)8-9-16(12)32-18(34)14-6-5-7-15(31)17(14)19(35)33-20(2,3)11-38(4,36)37/h5-10H,11H2,1-4H3,(H,32,34)(H,33,35) |
| SMILES | Cc1cc(ccc1NC(=O)c2cccc(I)c2C(=O)NC(C)(C)CS(=O)(=O)C)C(F)(C(F)(F)F)C(F)(F)F |
| IUPAC Name | 1-N-[4-(1,1,1,2,3,3,3-heptafluoropropan-2-yl)-2-methylphenyl]-3-iodo-2-N-(2-methyl-1-methylsulfonylpropan-2-yl)benzene-1,2-dicarboxamide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction