missing translation for 'onlineSavingsMsg'
Learn More

Flucloxacillin sodium, MedChemExpress

Product Code. 16428046
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16428046 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16428046 Supplier Medchem Express Supplier No. HYA0246A
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Flucloxacillin sodium is an active antibiotic against gram-positive and gram-negative bacteria.

Flucloxacillin sodium is an active antibiotic against gram-positive and gram-negative bacteria.

Specifications

Chemical Name or Material Flucloxacillin sodium
Physical Form Solid
CAS 1847-24-1
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C19H16ClFN3NaO5S
For Use With (Application) COVID-19-immunoregulation
Solubility Information H2O : 100 mg/mL (210.15 mM; Need ultrasonic) ∣DMSO : ≥ 100 mg/mL (210.15 mM)
SMILES O=C([C@@H]1N(C2=O)[C@]([C@@H]2NC(C(C(C(C(F)=CC=C3)=C3Cl)=NO4)=C4C)=O)([H])SC1(C)C)O[Na]
Molecular Weight (g/mol) 475.85
Formula Weight 475.85
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.