missing translation for 'onlineSavingsMsg'
Learn More

Fluconazole, TRC

Product Code. 30041228
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30041228 500 mg 500mg
30040429 100 mg 100mg
30040847 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30041228 Supplier Toronto Research Chemicals Supplier No. TRCF421000500MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86386-73-4
Molecular Formula C13 H12 F2 N6 O
Molecular Weight (g/mol) 306.27
Synonym 1H-1,2,4-Triazole-1-ethanol, α-(2,4-difluorophenyl)-α-(1H-1,2,4-triazol-1-ylmethyl)-, Biozolene, Diflucan, Elazor, Fluconazole, Flucostat, Flumycon, Flunazol, Flusol, Fluzon, Triflucan, UK 49858, Zoltec
IUPAC Name 2-(2,4-difluorophenyl)-1,3-bis(1,2,4-triazol-1-yl)propan-2-ol
SMILES OC(Cn1cncn1)(Cn2cncn2)c3ccc(F)cc3F

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C13H12F2N6O/c14-10-1-2-11(12(15)3-10)13(22,4-20-8-16-6-18-20)5-21-9-17-7-19-21/h1-3,6-9,22H,4-5H2
Formula Weight 306.1041
Percent Purity >95%
Chemical Name or Material Fluconazole
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.