missing translation for 'onlineSavingsMsg'
Learn More

Flucytosine, MedChemExpress

Product Code. 16452059
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16452059 1 g 1g
16430788 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16452059 Supplier Medchem Express Supplier No. HYB0139
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Flucytosine (5-Fluorocytosine, 5-FC, Ancobon), a fluorinated pyrimidine analogue, is an antifungal drug.

Flucytosine (5-Fluorocytosine, 5-FC, Ancobon), a fluorinated pyrimidine analogue, is an antifungal drug.

Specifications

Chemical Name or Material Flucytosine
Physical Form Solid
CAS 2022-85-7
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C4H4FN3O
For Use With (Application) COVID-19-immunoregulation
Synonym 5-Fluorocytosine NSC 103805 Ro 2-9915
Solubility Information DMSO : 16.67 mg/mL (129.13 mM; Need ultrasonic) ∣H2O : 6.67 mg/mL (51.67 mM; Need ultrasonic)
SMILES O=C1NC=C(F)C(N)=N1
Molecular Weight (g/mol) 129.09
Formula Weight 129.09
Percent Purity 99.77%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.