missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Fluo-4 NW Calcium Assay Kit

Product Code. 10266762
Change view
Click to view available options
Quantity:
10 Microplates
100 Microplate(s)
Unit Size:
1 set
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10266762 10 Microplates Each
10135132 100 Microplate(s) 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10266762 Supplier Molecular Probes™ Supplier No. F36206
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fluo-4 NW Calcium Assay Kit

Fluo-4 NW (No-Wash) Calcium Assay Kits offer a proprietary calcium assay formulation that requires neither a wash step nor a quencher dye. The fluo-4 NW assay achieves larger increases in fluorescence intensity than standard fluo-3 and fluo-4 assays with a wash step. Eliminating the wash step results in lower variability and higher Z' values than the standard fluo-4 assay, while providing an easier and faster assay as well.

The fluo-4NW indicator is nonfluorescent and stable in pH 7–7.5 buffer for several hours, so spontaneous conversion to the Ca2+-sensitive form is not a significant source of background fluorescence. Contributions to baseline fluorescence by the growth medium (e.g., esterase activity, proteins interacting with receptors of interest, or phenol red) are eliminated by removing the medium prior to adding the indicator dye to the wells.

Another source of potential fluorescence outside the cells is extrusion of the indicator out of the cell by organic anion transporters. Probenecid is commonly used to inhibit this transport and reduce the baseline signal. We have synthesized a proprietary water-soluble probenecid, which is supplied with the Fluo-4 NW Calcium Assay Kits. This form of probenecid has the advantages of being easy to dissolve in buffer and safer to use than the free acid, which requires caustic 1 M NaOH to dissolve. TheFluo-4 NW Calcium Assay Kits are designed for microplates and HTS, and the assay can be performed on adherent as well as nonadherent cells.

Fluo-4 AM is a fluorescent Ca+2 indicator that is widely used for in-cell measurement of agonist-stimulated and antagonist inhibited calcium signaling in high-throughput screening (HTS) applications. Its visible wavelength excitation (compatible with argon-ion laser sources), high sensitivity, and large fluorescence increase upon binding Ca2+ has made it the indicator of choice for characterizing G-protein–coupled receptor (GPCR) pharmacology and function. These properties have made fluo-4 AM attractive not only for microplate screening applications but for microscopy and flow cytometry as well.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Calcium Indicator
For Use With (Equipment) Microplate Reader
Product Type Stain
Dye Type Fluo-4NW
Quantity 10 Microplates
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.