missing translation for 'onlineSavingsMsg'
Learn More

Fluometuron, TRC

Product Code. 30408086
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30408086 1 g
30398599 5 g
30401791 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30408086 Supplier Toronto Research Chemicals Supplier No. TRCF5966501G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2164-17-2
Molecular Formula C10 H11 F3 N2 O
Molecular Weight (g/mol) 232.2
Synonym Urea, N,N-dimethyl-N'-[3-(trifluoromethyl)phenyl]-, Urea, 1,1-dimethyl-3-(α,α,α-trifluoro-m-tolyl)- (7CI,8CI), N,N-Dimethyl-N'-[3-(trifluoromethyl)phenyl]urea, 1,1-Dimethyl-3-(α,α,α-trifluoro-m-tolyl)urea, 3-(3-Trifluoromethylphenyl)-1,1-dimethylurea, 3-(m-Trifluoromethylphenyl)-1,1-dimethylurea, C 2059, Ciba 2059, Cotoran, Cotoran 85DF, Fluomethuron, Fluometuron, Lanex, N-(3-Trifluoromethylphenyl)-N',N'-dimethylurea, N-(m-Trifluoromethylphenyl)-N',N'-dimethylurea, Pakhtaran
IUPAC Name 1,1-dimethyl-3-[3-(trifluoromethyl)phenyl]urea
SMILES CN(C)C(=O)Nc1cccc(c1)C(F)(F)F

Specifications

Quantity 1 g
InChI Formula InChI=1S/C10H11F3N2O/c1-15(2)9(16)14-8-5-3-4-7(6-8)10(11,12)13/h3-6H,1-2H3,(H,14,16)
Formula Weight 232.0823
Chemical Name or Material Fluometuron
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.