missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Fluorescent Particle Array (Streptavidin, Odd # peaks) Kit

Product Code. 18664824 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18664824 5 x 1 mL 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18664824 Supplier Novus Biologicals™ Supplier No. NBP318598

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used to develop assays for allergy testing, autoimmune diseases, cardiac markers, cytokine detection, endocrine markers, infectious disease markers, isotyping, genotyping, kinase and phosphorylated protein activity, metabolic markers, and tissue typing

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 4°C. Do not freeze.
For Use With (Application) Flow Cytometry
Quantity 5 x 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.