missing translation for 'onlineSavingsMsg'
Learn More

Fluorobenzene, 99%

Product Code. 11904521 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11904521 2.5 kg Glass bottle 2.5kg
10733671 100 g Glass bottle 100g
10317040 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11904521 Supplier Thermo Scientific Acros Supplier No. 119340025
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 462-06-6 | C6H5F | 96.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 462-06-6
Molecular Formula C6H5F
Molecular Weight (g/mol) 96.10
MDL Number MFCD00000280
InChI Key PYLWMHQQBFSUBP-UHFFFAOYSA-N
Synonym benzene, fluoro, monofluorobenzene, phenyl fluoride, fluorbenzol, fluoro-benzene, 4-fluorobenzene, unii-g3tsz68k12, fluoranylbenzene, fluorobenzenes, 1,2,3,4,5-pentadeuterio-6-fluorobenzene
PubChem CID 10008
ChEBI CHEBI:5115
IUPAC Name fluorobenzene
SMILES FC1=CC=CC=C1

Specifications

Melting Point -42.0°C
Color Colorless
Density 1.0200g/mL
Boiling Point 85.0°C
Flash Point −15°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5F
Refractive Index 1.4643 to 1.4663
Quantity 2.5 kg
Beilstein 05,IV,632
Merck Index 15,4201
Specific Gravity 1.02
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol and water
Formula Weight 96.1
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Fluorobenzene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Fluorobenzene
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.