missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Fluotrac™ 600 High Binding Flat Bottomed Microplates

Product Code. 10525011
Change view
Click to view available options
Color:
Black, Clear Bottom
Unit Size:
Pack of 40
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10525011 Black, Clear Bottom Pack of 40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10525011 Supplier Greiner Bio-One Supplier No. 655097
  • ONLINE EXCLUSIVE PRICE
    266.00 €
    223.94 € / Pack of 40
    Save 42.06 €
    15% Off
 Restricted Item
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used for throughput screening.

  • Non-pyrogenic
  • Conical well design with flat bottom
  • Ensures accurate pipetting
  • Reduces dead volume of well
  • Free of detectable DNase, RNase, human DNA
  • Prevents wicking
TRUSTED_SUSTAINABILITY

Specifications

Barcode Alphanumeric
Certified Free of human DNA and pyrogens
Format Standard
Lid Without Lid
Array 8 x 12
Autoclavable Not Autoclavable
Binding Type High Binding
Color Black, Clear Bottom
Product Type Assay Microplate
Material Polystyrene
Volume (Metric) Well 25 to 340 μL
No. of Wells 96
Volume (Metric) Working 0.025 to 0.340 mL
Sterility Sterile
Well Shape Flat Bottom (F-well)
Dnase Free Yes
Max. RCF 4800 x g (swinging bucket rotor)
No. per Case 40
No. per Pack 10
Quantity 40/Cs.
RNase Free Yes
Temperature Range (Metric) -20°C to +60°C
Dimensions (L x W x H) 128 x 85 x 14 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.