missing translation for 'onlineSavingsMsg'
Learn More

Flupentixol dihydrochloride, MedChemExpress

Product Code. 16426878
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16426878 10 mg 10mg
16419417 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16426878 Supplier Medchem Express Supplier No. HY15856B
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Flupentixol is a high potency thioxanthene with D1 and D2 dopamine receptor antagonism. Flupentixol is used in therapy of schizophrenia as well as in anxiolytic and depressive disorders.

Flupentixol is a high potency thioxanthene with D1 and D2 dopamine receptor antagonism. Flupentixol is used in therapy of schizophrenia as well as in anxiolytic and depressive disorders.

Specifications

Chemical Name or Material Flupentixol dihydrochloride
Physical Form Solid
CAS 2413-38-9
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C23H27Cl2F3N2OS
For Use With (Application) Neuroscience-Neuromodulation
Synonym Flupenthixol dihydrochloride
Solubility Information H2O : 100 mg/mL (197.07 mM; Need ultrasonic) ∣DMSO : 33.33 mg/mL (65.68 mM; Need ultrasonic)
SMILES OCCN1CCN(CC/C=C2C3=C(SC4=C/2C=CC=C4)C=CC(C(F)(F)F)=C3)CC1.[H]Cl.[H]Cl
Molecular Weight (g/mol) 507.44
Formula Weight 507.44
Percent Purity 98.81%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.