missing translation for 'onlineSavingsMsg'
Learn More

Flupropanate, Dr. Ehrenstorfer

Product Code. 30315130
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30315130 25 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30315130 Supplier Dr. Ehrenstorfer Supplier No. DRECA13802000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Flupropanate
CAS 756-09-2
Analyte or Component Names Flupropanate
Shipping Condition +20°C
Molecular Formula C3 H2 F4 O2
Certifications/Compliance ISO/IEC 17025
InChI Formula InChI=1S/C3H2F4O2/c4-1(5)3(6,7)2(8)9/h1H,(H,8,9)
SMILES OC(=O)C(F)(F)C(F)F
IUPAC Name 2,2,3,3-tetrafluoropropanoic acid
Molecular Weight (g/mol) 146.04
Quantity 25 mg
Formula Weight 145.9991 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.