missing translation for 'onlineSavingsMsg'
Learn More

Fluralaner, MedChemExpress

Product Code. 16493247
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493247 50 mg 50mg
16489757 5 mg 5mg
16467206 100 mg 100mg
16407238 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16493247 Supplier Medchem Express Supplier No. HY16973
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Fluralaner (INN) is a systemic insecticide and acaricide Fluralaner through potent blockage of GABA and L-glutamate gated chloride channels.

Fluralaner (INN) is a systemic insecticide and acaricide Fluralaner through potent blockage of GABA and L-glutamate gated chloride channels.

Specifications

Chemical Name or Material Fluralaner
Physical Form Solid
CAS 864731-61-3
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C22H17Cl2F6N3O3
For Use With (Application) COVID-19-immunoregulation
Synonym A1443 AH252723
Solubility Information DMSO : 200 mg/mL (359.52 mM; Need ultrasonic)
SMILES O=C(NCC(NCC(F)(F)F)=O)C1=CC=C(C2=NOC(C(F)(F)F)(C3=CC(Cl)=CC(Cl)=C3)C2)C=C1C
Molecular Weight (g/mol) 556.29
Formula Weight 556.29
Percent Purity 99.57%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.