Learn More
Memmert™ Flush-Fit Sets with Air Slots for SF750
Product Code. 17508355 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
SF750 Sterilizer
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
17508355 SF750 Sterilizer Each
1 options
Product Code. 17508355 Supplier Memmert™ Supplier No. SF750B29742
Add to basket
Add to basket
Stainless steel frame covering gap between oven and wall opening.
  • Stainless steel
  • With air slots
  • Techincal clarification required
Specifications
For Use With (Equipment) SF750 Sterilizer
Includes Air Slots
Material Stainless Steel