Learn More
Memmert™ Flush-Fit Sets with Air Slots for SN75plus
Product Code. 17528775 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
SN75plus Sterilizer
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
17528775 SN75plus Sterilizer Each
1 options
Product Code. 17528775 Supplier Memmert™ Supplier No. SN75plusB29732
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Stainless steel frame covering gap between oven and wall opening.
  • Stainless steel
  • With air slots
  • Techincal clarification required
Specifications
For Use With (Equipment) SN75plus Sterilizer
Includes Air Slots
Material Stainless Steel