missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Flutamide

Product Code. 16647072 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16647072 25 g 25g
16637072 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16647072 Supplier Thermo Scientific Chemicals Supplier No. 466450250
 Restricted Item
Estimated Shipment: 15-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13311-84-7
Molecular Formula C11H11F3N2O3
Molecular Weight (g/mol) 276.22
MDL Number MFCD00072009
InChI Key MKXKFYHWDHIYRV-UHFFFAOYSA-N
IUPAC Name 2-methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propanamide
SMILES CC(C)C(=O)NC1=CC=C(C(=C1)C(F)(F)F)[N+]([O-])=O

Specifications

Alpha Vector FLUTAMIDE
Melting Point 110.0°C to 114.0°C
Color White to Yellow
Infrared Spectrum Conforms
Packaging glass bottle
Quantity 25 g
Solubility Information insoluble aq. soln.
Formula Weight 276.21
Physical Form Powder
Chemical Name or Material Flutamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.