missing translation for 'onlineSavingsMsg'
Learn More

FMC63 scFv Mouse, PE, Clone: Y45, AcroBiosystems

Product Code. 17101491 Shop All ACROBiosystems Products
Change view
Click to view available options
Quantity:
200 Tests
25 Tests
Unit Size:
200 tests
25 tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17101491 25 Tests 25 tests
17146321 200 Tests 200 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17101491 Supplier ACROBiosystems Supplier No. FM3HPY5325TESTS
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FMC63 scFv Monoclonal antibody specifically detects FMC63 scFv.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FMC63 scFv
Classification Monoclonal
Clone Y45
Conjugate PE
Host Species Human (HEK293)
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20°C, aviod light
Isotype IgG1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.