Learn More
FMC63 scFv Mouse, PE, Clone: Y45, AcroBiosystems
Mouse Monoclonal Antibody
Brand:  ACROBiosystems FM3-HPY53-200tests
 View more versions of this product
Product Code. 17146321
  • 2590.00 € / 200 tests
Description
Description
FMC63 scFv Monoclonal antibody specifically detects FMC63 scFv.
Specifications
Specifications
FMC63 scFv
Y45
Human (HEK293)
RUO
-20°C, aviod light
Monoclonal
PE
200 Tests
Primary
IgG1