missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FMNL2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
216.00 € - 539.00 €
Specifications
| Antigen | FMNL2 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
|
18419320
|
Novus Biologicals
NBP1-83901-25ul |
25 μL |
216.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18263646
|
Novus Biologicals
NBP1-83901 |
0.1 mL |
539.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FMNL2 Polyclonal specifically detects FMNL2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Spécification
| FMNL2 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| FHOD2, FLJ37546, formin homology 2 domain containing 2, Formin homology 2 domain-containing protein 2, formin-like 2, formin-like protein 2, KIAA1902 | |
| FMNL2 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 114793 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:ERVEELEENISHLSEKLQDTENEAMSKIVELEKQLMQRNKELDVVREIYKDANTQVHTLRKMVKEKEEAIQRQSTLEKKIHELEKQGTIKIQKKGDGDIAILPVVASGTLSMGSEVVAGNSVGP | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit