missing translation for 'onlineSavingsMsg'
Learn More

Fmoc N-Hydroxysuccinimide Ester, TRC

Product Code. 30007879
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Unit Size:
100g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007879 250 g 250g
30010777 100 g 100g
30007370 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007879 Supplier Toronto Research Chemicals Supplier No. TRCF634000250G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82911-69-1
Molecular Formula C19 H15 N O5
Molecular Weight (g/mol) 337.33
Synonym Carbonic Acid 2,5-Dioxo-1-pyrrolidinyl 9H-Fluoren-9-ylmethyl Ester, Fmoc-OSu, 1-[[(9H-Fluoren-9-ylmethoxy)carbonyl]oxy]-2,5-pyrrolidinedione, (Fluorenylmethoxycarbonyl)hydroxysuccinimide Ester, Succinimidyl (9-Fluorenyl)methyl Carbonate, N-(9H-Fluoren-9-ylmethoxycarbonyloxy)succinimide;
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 9H-fluoren-9-ylmethyl carbonate
SMILES O=C(OCC1c2ccccc2c3ccccc13)ON4C(=O)CCC4=O

Specifications

Quantity 250 g
InChI Formula InChI=1S/C19H15NO5/c21-17-9-10-18(22)20(17)25-19(23)24-11-16-14-7-3-1-5-12(14)13-6-2-4-8-15(13)16/h1-8,16H,9-11H2
Formula Weight 337.095
Chemical Name or Material Fmoc N-Hydroxysuccinimide Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.