missing translation for 'onlineSavingsMsg'
Learn More

FMOC-O-tert-Butyl-L-serine, 98%, Thermo Scientific™

Product Code. 10154680 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10154680 1 g Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10154680 Supplier Thermo Scientific Acros Supplier No. 265320010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 71989-33-8
Melting Point 127°C to 131°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Molecular Formula C22H24NO5
MDL Number MFCD00037127
Quantity 1 g
Specific Rotation Condition + 25.00 (24.00°C c=1,EtOAc)
Specific Rotation +24° to +26° (24°C, 589nm) (c=1, EtOAc)
Synonym fmoc-ser tbu-oh, fmoc-o-tert-butyl-l-serine, fmoc-ser t-bu-oh, fmoc-ser but, fmoc-o-tert.butyl-l-serine, fmoc-ser tbu, fmoc-ser but-oh, pubchem10035, fmoc-l-ser otbu oh, fmoc-l-ser tbu-oh
InChI Key REITVGIIZHFVGU-IBGZPJMESA-M
SMILES CC(C)(C)OC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C([O-])=O
IUPAC Name (2S)-3-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoate
Molecular Weight (g/mol) 382.44
PubChem CID 2724633
Formula Weight 383.45
Percent Purity ≥97.5%
Physical Form Amorphous Powder
Chemical Name or Material FMOC-O-tert-Butyl-L-serine, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.