missing translation for 'onlineSavingsMsg'
Learn More

FMOC-O-tert-Butyl-L-threonine, 98%, Thermo Scientific™

Product Code. 10696271 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10696271 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10696271 Supplier Thermo Scientific Acros Supplier No. 265330010
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 71989-35-0
Melting Point 131.0°C to 134.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C23H27NO5
Linear Formula FMOC-NHCH(COOH)CH(CH3)OC(CH3)3
MDL Number MFCD00077075
Quantity 1 g
Specific Rotation Condition + 16.00 (24.00°C c=1,EtOAc)
Specific Rotation + 16.00
Synonym fmoc-thr tbu-oh, fmoc-o-tert-butyl-l-threonine, fmoc-thr t-bu-oh, n-9-fluorenylmethoxycarbonyl-o-tert-butyl-l-threonine, fmoc-thr but, 2s,3r-3-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, n-9h-fluoren-9-yl methoxy carbonyl-o-tert-butyl-l-threonine, fmoc-thr but-oh, fmoc-o-tert.butyl-l-threonine, fmoc-thr otbu-oh
InChI Key LZOLWEQBVPVDPR-VLIAUNLRSA-N
SMILES C[C@@H](OC(C)(C)C)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
IUPAC Name (2S,3R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[(2-methylpropan-2-yl)oxy]butanoic acid
Molecular Weight (g/mol) 397.47
PubChem CID 6364643
Formula Weight 397.45
Percent Purity 98%
Physical Form Fine Fluffy Crystalline Powder
Chemical Name or Material FMOC-O-tert-Butyl-L-threonine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.