missing translation for 'onlineSavingsMsg'
Learn More

Fmoc-Ser(O-α-D-GalNAc(OAc)3)-OH, MedChemExpress

Product Code. 16480277
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16480277 50 mg 50mg
16476637 5 mg 5mg
16474946 100 mg 100mg
16473918 10 mg 10mg
16472906 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16480277 Supplier Medchem Express Supplier No. HY104004
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Fmoc-Ser(O-α-D-GalNAc(OAc)3)-OH is a drug for cancer.

Fmoc-Ser(O-α-D-GalNAc(OAc)3)-OH is a drug for cancer.

Specifications

Chemical Name or Material Fmoc-Ser(O-α-D-GalNAc(OAc)3)-OH
CAS 120173-57-1
Color White
Purity Grade Notes Research
Molecular Formula C32H36N2O13
Quantity 50 mg
Synonym Fmoc-Ser-(GalNAc(Ac)3-alpha-D)-OH Fmoc-Ser[GalNAc(Ac)3-α-D]-OH Fmoc-Ser(Ac3AcNH-α-Gal)-OH
Solubility Information DMSO : ≥ 100 mg/mL (152.29 mM)
SMILES O=C(N[C@H](C(O)=O)CO[C@@H](O[C@H](COC(C)=O)[C@H](OC(C)=O)[C@@H]1OC(C)=O)[C@@H]1NC(C)=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
Molecular Weight (g/mol) 656.63
Formula Weight 656.63
Percent Purity 98.06%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.