missing translation for 'onlineSavingsMsg'
Learn More

Folinic acid calcium salt pentahydrate, MedChemExpress

Product Code. 16498076
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498076 100 mg 100mg
16422006 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16498076 Supplier Medchem Express Supplier No. HYB0080100mg
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Folinic acid calcium salt pentahydrate (Leucovorin calcium salt pentahydrate) is a biological folic acid and is generally administered along with methotrexate (MTX) as a rescue agent to decrease MTX-induced toxicity.

Folinic acid calcium salt pentahydrate (Leucovorin calcium salt pentahydrate) is a biological folic acid and is generally administered along with methotrexate (MTX) as a rescue agent to decrease MTX-induced toxicity.

Specifications

Chemical Name or Material Folinic acid calcium salt pentahydrate
Physical Form Solid
CAS 6035-45-6
Color Light Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C20H31CaN7O12
For Use With (Application) Cancer-programmed cell death
Synonym Leucovorin calcium salt pentahydrate
Solubility Information H2O : 10 mg/mL (16.62 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble)
SMILES O=CN1C(C2=O)=C(NC(N)=N2)NCC1CNC3=CC=C(C(N[C@H](C([O-])=O)CCC([O-])=O)=O)C=C3.O.O.O.O.[Ca+2].O
Molecular Weight (g/mol) 601.58
Formula Weight 601.58
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.