missing translation for 'onlineSavingsMsg'
Learn More

Hartmann Larochette™ Foliodress™ Non-woven Surgical Face Masks

Product Code. 11720104
Change view
Click to view available options
Color:
Green
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
11720104 Green Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11720104 Supplier Hartmann Larochette™ Supplier No. 9921431
  • ONLINE EXCLUSIVE PRICE
    22.05 €
    8.02 € / Pack of 50
    Save 14.03 €
    63% Off
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-quality surgical masks made of hyoallergenic, non-woven materials. Hartmann Larochette™ Foliodress™ Non-woven Surgical Face Masks are ideal for single use in surgical areas, isolation wards, and intensive care departments.

  • High-quality surgical face masks according to EN 14683:2014 type II or type II-R.
  • Hypoallergenic non-woven materials, free from natural rubber latex.
  • High filtration performance and comfortable breathing resistance
  • Long integrated nose bar.
  • Fragrance free.
  • Durable.

Specifications

Product Type General Purpose Face Mask
Color Green
Sterility Non-sterile
Certifications/Compliance EN 14683:2014 type II or type II-R
Construction Non-woven
Disposable Single-use
Filter Efficiency Bacterial
For Use With (Application) For single-use in all surgical areas, as well as on isolation wards and intensive care departments.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.