missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOLR2 Rabbit anti-Human, Clone: 1, Novus Biologicals™
Rabbit Monoclonal Antibody
379.00 € - 552.00 €
Specifications
| Antigen | FOLR2 |
|---|---|
| Clone | 1 |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:2000-1:10000 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18205848
|
Novus Biologicals
NBP2-89846-50UL |
50 μg |
379.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18205507
|
Novus Biologicals
NBP2-89846-100UL |
100 μg |
552.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FOLR2 Monoclonal specifically detects FOLR2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| FOLR2 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:2000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 2350 | |
| IgG | |
| Protein A purified |
| 1 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| BETA-HFR, FBP, FBP/PL-1, Folate receptor 2, folate receptor 2 (fetal), folate receptor beta, folate receptor, beta, Folate receptor, fetal/placental, folate-binding protein, fetal/placental, FR-BETA, FR-P3, Placental folate-binding protein | |
| Recombinant Human FOLR2 protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title