missing translation for 'onlineSavingsMsg'
Learn More

Fomepizole, MedChemExpress

Product Code. 16491078
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491078 5 g 5g
16462289 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16491078 Supplier Medchem Express Supplier No. HYB0876
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Fomepizole (4-Methylpyrazole) is a potent cytochrome P450 (CYP2E1) inhibitor. Fomepizole is a competitive inhibitor of the enzyme alcohol dehydrogenase. Fomepizole blocks further conversion of methanol and ethylene glycol to toxic metabolites. Fomepizole has the potential for an antidote for ethylene glycol or methanol poisoning.

Fomepizole (4-Methylpyrazole) is a potent cytochrome P450 (CYP2E1) inhibitor. Fomepizole is a competitive inhibitor of the enzyme alcohol dehydrogenase. Fomepizole blocks further conversion of methanol and ethylene glycol to toxic metabolites. Fomepizole has the potential for an antidote for ethylene glycol or methanol poisoning.

Specifications

Chemical Name or Material Fomepizole
Physical Form Solid-Liquid Mixture
CAS 7554-65-6
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C4H6N2
Synonym 4-Methylpyrazole
Solubility Information H2O : 100 mg/mL (1218.03 mM; Need ultrasonic) ∣DMSO : ≥ 100 mg/mL (1218.03 mM)
SMILES CC1=CNN=C1
Molecular Weight (g/mol) 82.1
Formula Weight 82.1
Percent Purity 99.54%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.