missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde, Extra Pure, Solution 37-41%, SLR, Fisher Chemical™

Product Code. 10160052 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
25 L
Packaging:
HDPE plastic bottle
Plastic container
Unit Size:
1 L
2.5 L
25 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10160052 2.5 L HDPE plastic bottle 2.5 L
10630813 500 mL HDPE plastic bottle 500mL
10231622 1 L HDPE plastic bottle 1 L
10170052 25 L Plastic container 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10160052 Supplier Fisher Chemical Supplier No. F/1501/PB17
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CH2O, CAS Number-50-00-0, 2.5L, 97 deg.C, CHEBI:16842, Colorless, 30.02g/mol, WSFSSNUMVMOOMR-UHFFFAOYSA-N, formaldehyde, -15 deg.C, 3274, 30.026, HDPE plastic bottle, 3 to 4.2, Liquid, 712, C-O, > 1.0, 2mbar at 20 deg.C | CAS: 50-00-0 | CH2O | 30.03 g/mol

CH2O, CAS Number-50-00-0, 2.5L, 97 deg.C, CHEBI:16842, Colorless, 30.02g/mol, WSFSSNUMVMOOMR-UHFFFAOYSA-N, formaldehyde, -15 deg.C, 3274, 30.026, HDPE plastic bottle, 3 to 4.2, Liquid, 712, C-O, > 1.0, 2mbar at 20 deg.C

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 50-00-0
Molecular Formula CH2O
Molecular Weight (g/mol) 30.03
MDL Number MFCD00003274
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O

Especificaciones

Packaging HDPE plastic bottle
Vapor Pressure 2mbar at 20°C
Vapor Density > 1.0
Physical Form Liquid
Color Colorless
pH 3 to 4.2
Melting Point -15°C
Boiling Point 97°C
Quantity 2.5 L
Formula Weight 30.02g/mol
compliance-icons compliance-icons compliance-icons compliance-icons
Nombre del producto
  • Formaldehyde Solution 37%
Palabra de advertencia
  • Danger
Clasificación
  • Acute toxicity Category 3
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Germ cell mutagenicity Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after single exposure Category 1
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H317-May cause an allergic skin reaction.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
  • H318-Causes serious eye damage.
  • H370-Causes damage to organs.
  • H314-Causes severe skin burns and eye damage.
  • H350-May cause cancer.
  • H341-Suspected of causing genetic defects.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
Consejos de prudencia
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Otros peligros
  • MIXTURE LIST-Contain: Formaldehyde, Methanol
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.