missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde, Extra Pure, Solution 37-41%, SLR, Fisher Chemical™

Product Code. 10170052 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
25 L
Packaging:
HDPE plastic bottle
Plastic container
Unit Size:
1 L
2.5 L
25 L
500mL
Product Code. Quantity Packaging unitSize
10170052 25 L Plastic container 25 L
10630813 500 mL HDPE plastic bottle 500mL
10231622 1 L HDPE plastic bottle 1 L
10160052 2.5 L HDPE plastic bottle 2.5 L
4 options
This item is not returnable. View return policy

Product Code. 10170052

Brand: Fisher Chemical F/1501/25

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CH2O, CAS Number-50-00-0, 2.5L, 97 deg.C, CHEBI:16842, Colorless, 30.02g/mol, WSFSSNUMVMOOMR-UHFFFAOYSA-N, formaldehyde, -15 deg.C, 3274, 30.026, HDPE plastic bottle, 3 to 4.2, Liquid, 712, C-O, > 1.0, 2mbar at 20 deg.C | CAS: 50-00-0 | CH2O | 30.03 g/mol

CH2O, CAS Number-50-00-0, 2.5L, 97 deg.C, CHEBI:16842, Colorless, 30.02g/mol, WSFSSNUMVMOOMR-UHFFFAOYSA-N, formaldehyde, -15 deg.C, 3274, 30.026, HDPE plastic bottle, 3 to 4.2, Liquid, 712, C-O, > 1.0, 2mbar at 20 deg.C

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-00-0
Molecular Formula CH2O
Molecular Weight (g/mol) 30.03
MDL Number MFCD00003274
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O

Specifications

Packaging Plastic container
Vapor Pressure 2mbar at 20°C
Vapor Density > 1.0
Physical Form Liquid
Color Colorless
pH 3 to 4.2
Melting Point -15°C
Boiling Point 97°C
Quantity 25 L
Formula Weight 30.02g/mol
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Formaldehyde Solution 37%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
  • Specific target organ toxicity after single exposure Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H370-Causes damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.