missing translation for 'onlineSavingsMsg'
Learn More

Formononetin, MedChemExpress

Product Code. 16448888
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16448888 10 mg 10mg
16471496 200 mg 200mg
16422366 500 mg 500mg
16419026 100 mg 100mg
16404787 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16448888 Supplier Medchem Express Supplier No. HYN018310mg
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Formononetin is a potent FGFR2 inhibitor with an IC50 of ∼4.31 μM. Formononetin potently inhibits angiogenesis and tumor growth.

Formononetin is a potent FGFR2 inhibitor with an IC50 of ∼4.31 μM. Formononetin potently inhibits angiogenesis and tumor growth.

Specifications

Chemical Name or Material Formononetin
CAS 485-72-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C16H12O4
Quantity 10 mg
Synonym Biochanin B Flavosil Formononetol
Solubility Information DMSO : ≥ 35 mg/mL (130.47 mM)
SMILES O=C1C(C2=CC=C(OC)C=C2)=COC3=CC(O)=CC=C13
Molecular Weight (g/mol) 268.26
Formula Weight 268.26
Percent Purity 97.67%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.