missing translation for 'onlineSavingsMsg'
Learn More

Fosbretabulin disodium, MedChemExpress

Product Code. 16467348
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16467348 10 mg 10mg
16429837 5 mg 5mg
16473367 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16467348 Supplier Medchem Express Supplier No. HY17449

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Fosbretabulin disodium (CA 4DP) is a tubulin destabilizing agent. Fosbretabulin disodium is the Combretastatin A4 prodrug that selectively targets endothelial cells, induces regression of nascent tumour neovessels, reduces tumour blood flow and causes central tumour necrosis.

Fosbretabulin disodium (CA 4DP) is a tubulin destabilizing agent. Fosbretabulin disodium is the Combretastatin A4 prodrug that selectively targets endothelial cells, induces regression of nascent tumour neovessels, reduces tumour blood flow and causes central tumour necrosis.

Specifications

Chemical Name or Material Fosbretabulin disodium
CAS 168555-66-6
Color White
Purity Grade Notes Research
Molecular Formula C18H19Na2O8P
Quantity 10 mg
Synonym CA 4DP CA 4P Combretastatin A4 disodium phosphate
Solubility Information H2O : 4.5 mg/mL (10.22 mM; Need ultrasonic and warming) ∣DMF : 1 mg/mL (2.27 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble)
SMILES COC1=C(OC)C(OC)=CC(/C=C\C2=CC=C(OC)C(OP(O[Na])(O[Na])=O)=C2)=C1
Molecular Weight (g/mol) 440.29
Formula Weight 440.29
Percent Purity 98.06%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.