missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fosbretabulin disodium, MedChemExpress
Description
Fosbretabulin disodium (CA 4DP) is a tubulin destabilizing agent. Fosbretabulin disodium is the Combretastatin A4 prodrug that selectively targets endothelial cells, induces regression of nascent tumour neovessels, reduces tumour blood flow and causes central tumour necrosis.
Specifications
Specifications
| Chemical Name or Material | Fosbretabulin disodium |
| CAS | 168555-66-6 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C18H19Na2O8P |
| Quantity | 50 mg |
| Synonym | CA 4DP CA 4P Combretastatin A4 disodium phosphate |
| Solubility Information | H2O : 4.5 mg/mL (10.22 mM; Need ultrasonic and warming) ∣DMF : 1 mg/mL (2.27 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble) |
| SMILES | COC1=C(OC)C(OC)=CC(/C=C\C2=CC=C(OC)C(OP(O[Na])(O[Na])=O)=C2)=C1 |
| Molecular Weight (g/mol) | 440.29 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?