missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FOXA3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106980
This item is not returnable.
View return policy
Description
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 92%; Human: 100%; Mouse: 92%; Rabbit: 85%; Rat: 100%; Zebrafish: 79% Immunogen sequence: FENGCYLRRQ KRFKLEEKVK KGGSGAATTT RNGTGSAAST TTPAATVTSP.
FOXA3 encodes a member of the forkhead class of DNA-binding proteins. These hepatocyte nuclear factors are transcriptional activators for liver-specific transcripts such as albumin and transthyretin, and they also interact with chromatin. Similar family members in mice have roles in the regulation of metabolism and in the differentiation of the pancreas and liver. The crystal structure of a similar protein in rat has been resolved.
Specifications
| FOXA3 | |
| Polyclonal | |
| Unconjugated | |
| FOXA3 | |
| FKHH3; Fork head-related protein FKH H3; forkhead box A3; forkhead box protein A3; Foxa3; hepatocyte nuclear factor 3 gamma (winged helix transcription factor); hepatocyte nuclear factor 3, gamma; hepatocyte nuclear factor 3-gamma; HNF3G; HNF-3G; HNF-3-gamma; Tcf3g; Tcf-3g; Transcription factor 3G | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 15377, 3171 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| P35584, P55318 | |
| FOXA3 | |
| Synthetic peptide directed towards the middle region of human FOXA3 (aa 201-250). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction