missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXL2 Antibody (CL12753), Novus Biologicals™
Mouse Monoclonal Antibody
401.00 € - 649.00 €
Specifications
| Antigen | FOXL2 |
|---|---|
| Clone | CL12753 |
| Dilution | Western Blot 1 μg/ml, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18671665
|
Novus Biologicals
NBP3-18569-25ul |
25 μg |
401.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18650494
|
Novus Biologicals
NBP3-18569-100ul |
100 μg |
649.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FOXL2 Monoclonal antibody specifically detects FOXL2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| FOXL2 | |
| Western Blot 1 μg/ml, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BPES, BPES1PINTO, forkhead box L2, forkhead box protein L2, forkhead transcription factor FOXL2, PFRK, POF3 | |
| This antibody was developed against a sythetic peptide for P58012. Actual immunogen sequence is proprietary. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| CL12753 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Epigenetics | |
| PBS, pH 7.2, 40% glycerol | |
| 668 | |
| IgG3 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title