missing translation for 'onlineSavingsMsg'
Learn More

FOXL2 Antibody (CL12762), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-18544-100ul

 View more versions of this product

Product Code. 18640195

  • 685.00 € / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

FOXL2 Monoclonal antibody specifically detects FOXL2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FOXL2
Monoclonal
Unconjugated
PBS, pH 7.2, 40% glycerol
Mouse
Protein A purified
RUO
Primary
Human
Purified
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL12762
Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
BPES, BPES1PINTO, forkhead box L2, forkhead box protein L2, forkhead transcription factor FOXL2, PFRK, POF3
This antibody was developed against a sythetic peptide for P58012. Actual immunogen sequence is proprietary.
100 μg
Epigenetics
668
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG3
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.