Learn More
FoxO1/FKHR Antibody (CL13278) [HRP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-18557H
Product Code. 30303177
  • 770.00 € / 0.10mL
Description
FoxO1/FKHR Monoclonal antibody specifically detects FoxO1/FKHR in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FoxO1/FKHR
Monoclonal
HRP
FKH1, FKHRforkhead box protein O1, forkhead box O1, Forkhead box protein O1A, forkhead homolog in rhabdomyosarcoma, Forkhead in rhabdomyosarcoma, FOXO1Aforkhead, Drosophila, homolog of, in rhabdomyosarcoma
This antibody was developed using a synthetic peptide derived from Q12778, with the exact immunogen sequence remaining proprietary.
0.1 mL
Apoptosis, Cancer, DNA Repair, Growth and Development, Neuronal Cell Markers, Phospho Specific
2308
Store at 4⪚C in the dark.
IgG1
Western Blot
CL13278
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified