Learn More
FoxO1/FKHR, p Ser256 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand:  Novus Biologicals NB100-81927
 View more versions of this product
Product Code. 18711614
  • 436.00 € / 0.05mL
Description
FoxO1/FKHR Polyclonal specifically detects FoxO1/FKHR in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.