Learn More
FOXO3 Antibody (CL13589) [Alexa Fluor™ 700], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-24564AF700
Product Code. 30303716
  • 737.00 € / 0.10mL
Description
FOXO3 Monoclonal antibody specifically detects FOXO3 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FOXO3
Monoclonal
Alexa Fluor 700
AF6q21, AF6q21 protein, DKFZp781A0677, FKHRL1MGC31925, FKHRL1P2, forkhead box O3, forkhead box protein O3, forkhead homolog (rhabdomyosarcoma) like 1, Forkhead in rhabdomyosarcoma-like 1, forkhead, Drosophila, homolog of, in rhabdomyosarcoma-like 1, FOXO2, FOXO3Aforkhead box O3A, MGC12739
This antibody was developed using a synthetic peptide derived from O43524, with the exact immunogen sequence remaining proprietary.
0.1 mL
Apoptosis, Cancer, DNA Repair, Phospho Specific, Transcription Factors and Regulators
2309
Store at 4⪚C in the dark.
IgG2a
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL13589
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified