missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXO4 Recombinant Rabbit Monoclonal Antibody (23GB1470), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen MA553995
This item is not returnable.
View return policy
Description
FOXO4 Recombinant Monoclonal antibody specifically detects FOXO4 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot
Specifications
| FOXO4 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| FOXO4 | |
| AFX; Afx1; Afxh; AFXzeta; Fkhr3; Fork head domain transcription factor AFX1; forkhead box O4; forkhead box protein O4; forkhead protein; FOXO4; FOXO4b; MGC120490; MLLT7; myeloid/lymphoid or mixed lineage-leukemia translocation to 7 homolog; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 7; OTTHUMP00000023497; RGD1561201 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 302415, 4303, 54601 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB1470 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P98177, Q9WVH3 | |
| FOXO4 | |
| A synthesized peptide derived from human FoxO4 (450-505AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction