missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FoxP1 Antibody (OTI4F1), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71831PE
This item is not returnable.
View return policy
Description
FoxP1 Monoclonal antibody specifically detects FoxP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| FoxP1 | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| OTI4F1 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
| FLJ23741, fork head-related protein like B, forkhead box P1, forkhead box protein P1, glutamine-rich factor 1, hFKH1B, HSPC215, MGC12942, MGC88572, MGC99551, QRF112CC4 | |
| Full length human recombinant protein of human FOXP1(NP_116071) produced in HEK293T cell. | |
| 0.1 mL | |
| Cancer | |
| 27086 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction