missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FoxP3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 24 publications
Brand: Novus Biologicals NB100-39002SS
This item is not returnable.
View return policy
Description
FoxP3 Polyclonal specifically detects FoxP3 in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting, Single Cell Western, Dual RNAscope ISH-IHC.
Specifications
| FoxP3 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| AIID, AIIDMGC141961, anti-human foxp3, anti-mouse foxp3, DIETER, forkhead box P3, Forkhead Box Protein P3, foxp3 ihc, foxp3 paraffin, FOXP3delta7, immune dysregulation, polyendocrinopathy, enteropathy, X-linked, Immunodeficiency, Polyendocrinopathy, Enteropathy, X-Linked, IPEX, JM2, MGC141961, MGC141963, PIDX, PIDXMGC141963, scurfin, XPID, XPIDpolyendocrinopathy, enteropathy, X-linked | |
| Rabbit | |
| Epitope affinity purified | |
| RUO | |
| Primary | |
| Mouse. This antibody shows reactivity against mouse CD4+CD25+T cells. There appears to be no reactivity against human Jurkat and mouse CD4+CD25-T cell lysates. Human reactivity reported in scientific literature (PMID: 23922187). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:2000, Flow Cytometry 1:8000, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation, Immunohistochemistry-Paraffin 1:400. Use reported by customer review, Immunoblotting reported in scientific literature (PMID 26485285), Single Cell Western 100 ug/mL, Dual RNAscope ISH-IHC | |
| Q99JB6 | |
| FOXP3 | |
| A peptide corresponding to amino acids 43-100 of mouse FOXP3. [Swiss-Prot# Q99JB6] | |
| 0.025 mL | |
| Adaptive Immunity, Cell Biology, Chromatin Research, Immunology, Regulatory Immunology, Transcription Factors and Regulators | |
| 50943 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction