Learn More
FoxP3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 24 publications
Brand: Novus Biologicals NB100-39002
This item is not returnable.
View return policy
Description
FoxP3 Polyclonal specifically detects FoxP3 in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting, Single Cell Western, Dual RNAscope ISH-IHC.
Specifications
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| Rabbit | |
| Purified |
| Polyclonal | |
| 0.1 mL | |
| IgG |