missing translation for 'onlineSavingsMsg'
Learn More

Foxp3 Mouse, FITC, Clone: 3G3, enQuireBio™

Mouse Monoclonal Antibody

248.85 € - 459.90 €

Specifications

Antigen Foxp3
Clone 3G3
Concentration 0.2mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15972398
View Documents
EnquireBio
QAB71-F-25ug
25 μg
248.85 €
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15962398
View Documents
EnquireBio
QAB71-F-100ug
100 μg
459.90 €
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

Foxp3
0.2mg/mL
Monoclonal
Mouse
10 mM NaH2PO4, 150 mM NaCl, 0.05% BSA, 0.05% NaN3, pH7.2
20371
IgG1 κ
Protein A/G Purified
3G3
Flow Cytometry
FITC
RUO
NP_001186276
Foxp3
Primary
Store at 2-4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.