Learn More
Invitrogen™ FPR2 Recombinant Rabbit Monoclonal Antibody (HL2664)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA556716
Product Code. 30276478
  • 500.00 € / 100µL
Description
FPR2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Formyl peptide receptor 2 (FPR2) is a low affinity receptor for N-formyl-methionyl peptides. Binding of FMLP to FPR2 causes activation of neutrophils. The response is mediated via a G-protein that activates a phosphatidylinositol-calcium second messenger system. The activation of LXA4R could result in an anti-inflammatory outcome counteracting the actions of proinflammatory signals such as LTB4.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FPR2
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
O88536, P25090
FPR2
Recombinant protein encompassing a sequence within the Extracellular domain of human FPRL1. The exact sequence is proprietary.
100 μL
Primary
Human, Mouse
Antibody
IgG
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
HL2664
Unconjugated
FPR2
ALXR; E330010I07Rik; fMLPR1; FMLP-related receptor I; FMLP-R-I; FMLP-R-II; FMLPX; formyl peptide receptor 2; formyl peptide receptor, related sequence 2; formyl peptide receptor, related sequence 2-like; formyl peptide receptor-like 1; Formylpeptide receptor-related sequence 2; FPR2; FPR2A; FPRH1; FPRH2; Fprl1; Fprl-1; FPR-like 1; Fpr-rs1; Fpr-rs2; HM63; Lipoxin A4 receptor; lipoxin A4 receptor (formyl peptide receptor related); lipoxin A4 receptor-like protein; Lipoxin receptor; LXA4 receptor; LXA4R; N-formyl peptide receptor 2; N-formylpeptide receptor-like 2; RFP
Rabbit
Protein A
RUO
14289, 2358
-20°C, Avoid Freeze/Thaw Cycles