Learn More
FPRL1/FPR2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 21 publication
Brand:  Novus Biologicals NLS1878
 View more versions of this product
Product Code. 18797423
  • 595.00 € / 0.05mL
Description
FPRL1/FPR2 Polyclonal specifically detects FPRL1/FPR2 in Human, Mouse, Rat, Bacteria samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Electron Microscopy.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Immunofluorescence, Immunohistochemistry (Paraffin)
Western Blot 1:1000, Flow Cytometry, Immunohistochemistry 1:300, Immunocytochemistry/Immunofluorescence 1:20-1:75, Immunohistochemistry-Paraffin 1:300, Immunohistochemistry-Frozen reported in scientific literature (PMID 24086560), Electron Microscopy
FPR2
Affinity Purified
Polyclonal
ALXR, FMLP-R-II, FMLPX, formyl peptide receptor 2, Formyl peptide receptor-like 1RFP, FPR2A, FPRH1FMLP-related receptor I, FPRL1LXA4 receptor, HM63FPRH2, Lipoxin A4 receptor, lipoxin A4 receptor (formyl peptide receptor related), LXA4RFMLP-R-I, N-formyl peptide receptor 2
38 kDa