missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Frames and Accessories for Immuno Breakable Modules, BreakApart frame
BIOPHARMA

Product Code. 10585211
Change view
Click to view available options
Product Type:
Frame
No. per Pack:
5
50
No. per Case:
50
60
Plate Compatibility:
Immuno Standard Modules
Nunc C8 Breakable Modules
Unit Size:
Pack of 25
Pack of 60
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type No. per Pack No. per Case Plate Compatibility Unit Size
10585211 Frame 5 60 Nunc C8 Breakable Modules Pack of 60
10524641 Frame 5 60 Immuno Standard Modules Pack of 60
10733316 Frame 50 50 Nunc C8 Breakable Modules Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10585211 Supplier Thermo Scientific™ Supplier No. 431615
  • ONLINE EXCLUSIVE PRICE
    95.05 €
    90.59 € / Pack of 60
    Save 4.46 €
    4% Off
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use Thermo Scientific™ Frames and Accessories for Immuno Breakable Modules with Nunc™ C8 Breakable Modules.

Use Thermo Scientific™ Frames and Accessories for Immuno Breakable Modules with Nunc™ C8 Breakable Modules. These frames include tools to facilitate removing the strips.

Warranty: 90 days
TRUSTED_SUSTAINABILITY

Specifications

Format Strip Plate
Description High optical quality for reliable results
Array 8 x 12
Packaging 60 Cases, 5 Pk.
Color White
Product Type Frame
Material Polystyrene
No. of Wells 96
Well Shape Curved Edges, Flat Bottom (C-well)
For Use With (Application) For solid phase immuno assays
No. per Case 60
No. per Pack 5
Plate Compatibility Nunc C8 Breakable Modules
Show More Show Less
Which 96-well Immuno plates or modules do you recommend for various applications?

The following list offers a brief description of the features of Nunc products and their specific applications:
- Nunc-Immuno Plates, MaxiSorp surface - These plates are designed for solid phase immuno assays and have a polystyrene surface with high affinity for polar groups and hydrophilic molecules. These 96-well plates are available with flat (F), round (U), or (C) bottom-well designs.
- Nunc-Immuno Plates, PolySorp surface - These plates have a polystyrene surface which adsorps less polar molecules compared to the MaxiSorp surface and has a high affinity for hydrophobic groups. These 96-well plates are available with flat (F), round (U), or (C) bottom-well designs.
- Nunc-Immuno Modules - These modules are designed for solid-phase immuno assays. The modules are available in 8-, 12- or 16-well formats with (F), (U), and (C) bottom-well and 8-well BreakApart with (C) bottom-well designs. These different formats allow one to choose the style which is appropriate for their assay design.
BreakApart Modules and LockWell Modules can be used for radioimmunoassays.
- Nunc StarWell Immuno Breakable Modules - The 8-well modules feature eight fins on the inner wall of the (C) bottom wells. This design increases the surface area by 50%. The increase in surface area allows more molecules to be immobilized, increasing assay signal. The fin configuration provides shorter diffusion distance to the surface, reducing incubation times.
- LockWell Immuno Breakable Modules - Each plate consists of 1 x 8 breakable strips. These strips are assembled in a designed frame which locks each well into place by a spring lock. This spring lock design orientates each well at the same horizontal level allowing uniform washing and reading. The LockWell Modules are available with round (U), (C) or StarWell bottom-well designs.
- Plates and Modules with (CovaLink) Covalent Binding Surfaces: These plates can be used to bind proteins, peptides, DNA or carbohydrates. Covalent linkage occurs via secondary amine group. This allows a specific orientation and provides improved stability compared to passive adsorption due to chemical binding between molecules. Binding of molecules on PolySorp and MaxiSorp surfaces is obtained by passive adsorption. CovaLink is a surface grafted with secondary amino groups (<NH) which serves as a bridge for covalent coupling. The covalent binding can immobilize small molecules such as biotin or peptides which may otherwise bind weakly by physical adsorption and facilitate the recognition by detection molecule.
- Nunc FluoroNunc/LumiNunc 96-Well Plates: These plates are optimized for IFMA (Immunofluorometric Assays) or FIA (Fluorometric Immuno Assays). The transparent polystyrene plates give a low background fluorescence and are optimal where a read-through system is used. The white plates/modules provide maximum reflection of fluorescence signal while maintaining low background. The white plates/modules are often used for epifluorescence reading. Black modules reduce background fluorescence and minimize back-scatter light which is often encountered in epifluorescence.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.