missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Frizzled-2 Polyclonal Antibody

Description
Percent identity with other species by BLAST analysis: Human, Mouse, Rat, Hamster, Elephant, Panda, Dog, Bovine, Horse, Pig (100%) Sheep, Rabbit (94%) Opossum (89%).
This intronless gene is a member of the frizzled gene family. Members of this family encode seven-transmembrane domain proteins that are receptors for the wingless type MMTV integration site family of signaling proteins. This gene encodes a protein that is coupled to the beta-catenin canonical signaling pathway. Competition between the wingless-type MMTV integration site family, member 3A and wingless-type MMTV integration site family, member 5A gene products for binding of this protein is thought to regulate the beta-catenin-dependent and -independent pathways.
Specifications
Specifications
| Antigen | Frizzled-2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Gene | Fzd2 |
| Gene Accession No. | Q08464, Q14332, Q9JIP6 |
| Gene Alias | AL033370; AW456835; Dfrizzled2; Drosophila polarity gene (frizzled) homologue; Drosophila polarity gene homolog 2; frizzled 2, seven transmembrane spanning receptor; frizzled class receptor 2; frizzled family receptor 2; frizzled homolog 2; frizzled homolog 2 (Drosophila); frizzled-10; Frizzled-2; Fz10; fz-10; Fz2; Fz-2; Fzd10; Fzd2; fzE2; hFz2; Mfz10; Mfz10a; mFz2; rFz2 |
| Gene Symbols | Fzd2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?