missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Frizzled-9 Antibody (vantictumab) - IgG2SA, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28265-50ug
This item is not returnable.
View return policy
Description
Frizzled-9 Monoclonal antibody specifically detects Frizzled-9 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Frizzled-9 | |
| Monoclonal | |
| Unconjugated | |
| CD349, CD349 antigen, frizzled (Drosophila) homolog 9, frizzled homolog 9 (Drosophila), frizzled-9, fz-9, FZD3fzE6, FzE6, hFz9 | |
| FZD | |
| 50 μg | |
| GPCR, Growth and Development, Neuronal Cell Markers, Signal Transduction, Wnt Signaling Pathway | |
| 8326 | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| vantictumab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction